3,4-Dimethoxy-2-methylpyridine 97%

Supplier: Ambeed
Warning

A235442-1G A235442-5G A235442-100MG A235442-250MG
103811-016EA 39 USD
103811-016 103850-784 77089-790 77089-792 77261-622 77261-624
3,4-Dimethoxy-2-methylpyridine 97%
3,4-Dimethoxy-2-methylpyridine

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD16610899
CAS Number: 107512-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR