(3,4-Dihydroxyphenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: DOPAC, 3,4-Dihydroxybenzeneacetic acid, 3,4-Dihydroxyphenylacetic acid

A115979-100G A115979-10G A115979-25G A115979-500G A115979-5G A115979-1G
103824-518EA 13 USD
103824-518 77245-918 77245-920 77245-922 77245-924 77311-340 77566-094
(3,4-Dihydroxyphenyl)acetic acid 98%
(3,4-Dihydroxyphenyl)acetic acid

Precaución: For research use only.

Formula: (C₆H₃(OH)₂)CH₂COOH
Melting Pt: 127…130 °C
Flash Pt: 221 °C
Storage Temperature: Ambient
MDL Number: MFCD00004338
CAS Number: 102-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR