3,4-Dihydroxybenzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Protocatechuic aldehyde, Protocatechualdehyde

A152308-100G A152308-10G A152308-1G A152308-500G A152308-25G A152308-5G
103842-540EA 6 USD
103842-540 77150-418 77150-420 77150-422 77150-424 77299-940 77318-154
3,4-Dihydroxybenzaldehyde 98%
3,4-Dihydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₆O₃
MW: 138.12 g/mol
Melting Pt: 150…157 °C
Flash Pt: 146.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00003370
CAS Number: 139-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR