3,4-Dihydroisoquinolin-1(2H)-one 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A248375-100MG A248375-250MG A248375-1G A248375-25G A248375-5G A248375-10G
103811-822EA 12 USD
103811-822 103851-656 77013-938 77013-940 77090-998 77091-000 77304-446 77683-743
3,4-Dihydroisoquinolin-1(2H)-one 98%, Ambeed.Inc
3,4-Dihydroisoquinolin-1(2H)-one

Precaución: For research use only.

Formula: C9H9NO
Boiling Pt: 388.1 °C (760 mm Hg)
Melting Pt: 64…66 °C
Flash Pt: 226.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00853963
CAS Number: 1196-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR