5,6-Dihydro-4h-pyran-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dihydro-2H-pyran-6-carboxylic acid

A107135-100MG A107135-250MG
103797-130EA 18 USD
103797-130 103822-890 76765-538 77623-892
5,6-Dihydro-4h-pyran-2-carboxylic acid 95%
5,6-Dihydro-4h-pyran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₈O₃
MW: 128.126 g/mol
Boiling Pt: 271.4 °C (760 mm Hg)
Flash Pt: 119.1 °C
Storage Temperature: Freezer
MDL Number: MFCD08437234
CAS Number: 31518-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR