3,4-Dihydro-2H-1,4-benzothiazine 97%

Supplier: Ambeed
Warning

A357769-5G A357769-250MG A357769-1G
103826-200EA 33 USD
103826-200 103837-204 76840-364 77278-244 77306-686
3,4-Dihydro-2H-1,4-benzothiazine 97%
3,4-Dihydro-2H-1,4-benzothiazine

Precaución: for Research Use Only

Formula: C₈H₉NS
Melting Pt: 33…37 °C
Flash Pt: 119.8 °C
Storage Temperature: Ambient
MDL Number: MFCD04038593
CAS Number: 3080-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR