3,4-Dihydro-2H-benzo[b][1,4]oxazine-6-carbonitrile 95%

Supplier: Ambeed
Warning

A188865-100MG A188865-250MG A188865-1G
103816-138EA 483 USD
103816-138 103846-732 77274-376 77274-378 77302-154
3,4-Dihydro-2H-benzo[b][1,4]oxazine-6-carbonitrile 95%
3,4-Dihydro-2H-benzo[b][1,4]oxazine-6-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08544343
CAS Number: 928118-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR