4,5-Difluoro-2-nitroaniline 98%

Supplier: Ambeed
Warning

A114203-100G A114203-10G A114203-25G A114203-1G
103798-370EA 5 USD
103798-370 103824-210 77284-350 77284-352 77284-354 77297-298
4,5-Difluoro-2-nitroaniline 98%
4,5-Difluoro-2-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄F₂N₂O₂
Flash Pt: 140.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00010876
CAS Number: 78056-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR