3,4-Diethoxy-3-cyclobutene-1,2-dione 98%
Supplier: Ambeed
|
Danger
|
A112729-10G
A112729-25G
A112729-500G
A112729-1G
A112729-5G
A112729-100G
103798-092EA
5
USD
103798-092
103823-930
77189-160
77189-162
77189-164
77297-206
77310-752
77745-836
3,4-Diethoxy-3-cyclobutene-1,2-dione 98%
3,4-Diethoxy-3-cyclobutene-1,2-dione
Precaución: for Research Use Only
Formula:
C₈H₁₀O₄ Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001333 CAS Number: 5231-87-8 UN: 3082 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...