3,4-Dichlorophenylhydrazinium chloride 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dichlorophenylhydrazine hydrochloride, 1-(3,4-Dichlorophenyl)hydrazine hydrochloride

A195716-25G A195716-5G A195716-100G A195716-1G
103816-776EA 9 USD
103816-776 103847-388 77085-338 77085-340 77111-976 77302-478
3,4-Dichlorophenylhydrazinium chloride 98%
3,4-Dichlorophenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
Boiling Pt: 288.3 °C (760 mm Hg)
Melting Pt: 230 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012937
CAS Number: 19763-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR