3,4-Dichlorophenylboronic acid 98%

Supplier: Ambeed
Warning

A153220-25G A153220-5G A153220-100G A153220-10G A153220-500G A153220-1000G
103842-648EA 9 USD
103842-648 77318-288 77318-290 77611-446 77611-448 77685-266 77901-454
3,4-Dichlorophenylboronic acid 98%
3,4-Dichlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BCl₂O₂
Boiling Pt: 339.2 °C (760 mm Hg)
Melting Pt: 280…285 °C
Flash Pt: 159 °C
Storage Temperature: Ambient
MDL Number: MFCD01074646
CAS Number: 151169-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR