3,4-Dichloro-N-methylaniline 98%

Supplier: Ambeed
Warning

Synonyms: N1-Methyl-3,4-dichloroaniline

A857067-1G A857067-25G A857067-100G A857067-5G
103841-254EA 39 USD
103841-254 76845-512 76845-514 77281-734 77308-458
3,4-Dichloro-N-methylaniline 98%
3,4-Dichloro-N-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇Cl₂N
MW: 176.04 g/mol
Boiling Pt: 287.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00052850
CAS Number: 40750-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR