3,4-Dichloroisothiazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A128702-250MG A128702-5G A128702-1G A128702-25G
103801-174EA 39 USD
103801-174 77271-904 77298-318 77313-524 77313-526
3,4-Dichloroisothiazole-5-carboxylic acid 97%
3,4-Dichloroisothiazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₄HCl₂NO₂S
MW: 198.027 g/mol
Boiling Pt: 205.9 °C (760 mm Hg)
Melting Pt: 175…177 °C
Flash Pt: 78.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03789232
CAS Number: 18480-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR