3,4-Dichloro-2-fluoroaniline 98%

Supplier: Ambeed
Warning

A111713-100G A111713-25G A111713-1G A111713-5G
103797-892EA 9 USD
103797-892 103823-710 77108-000 77108-002 77284-170 77310-522
3,4-Dichloro-2-fluoroaniline 98%
3,4-Dichloro-2-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄Cl₂FN
MW: 180.007 g/mol
Boiling Pt: 255.8 °C (760 mm Hg)
Melting Pt: 57…61 °C
Flash Pt: 108.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039241
CAS Number: 886762-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR