3,4-Dibromothiophene 98%

Supplier: Ambeed
Warning

A404368-25G A404368-100G A404368-10G A404368-1G A404368-500G A404368-1000G A404368-5G
103810-290EA 187 USD
103810-290 103838-148 76841-630 77291-206 77291-208 77291-210 77291-212 77307-090 77307-092
3,4-Dibromothiophene 98%
3,4-Dibromothiophene

Precaución: for Research Use Only

Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 221.5 °C (760 mm Hg)
Melting Pt: 4…5 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005465
CAS Number: 3141-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR