3,4-Dibromopyridine 97%

Supplier: Ambeed
Warning

A143654-100G A143654-10G A143654-25G A143654-5G
103803-306EA 5 USD
103803-306 76996-986 76996-988 77316-596 77316-598
3,4-Dibromopyridine 97%
3,4-Dibromopyridine

Precaución: for Research Use Only

Formula: C₅H₃Br₂N
Boiling Pt: 239.9 °C (760 mm Hg)
Melting Pt: 71…72 °C
Flash Pt: 98.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00234016
CAS Number: 13534-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR