3,4-Dibromo-1H-pyrrole-2,5-dione 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dibromomaleimide

A249093-1G A249093-100G A249093-10G A249093-250MG
103811-856EA 12 USD
103811-856 103851-694 77091-038 77238-232 77238-234 77238-236
3,4-Dibromo-1H-pyrrole-2,5-dione 98%
3,4-Dibromo-1H-pyrrole-2,5-dione

Precaución: for Research Use Only

Formula: C4HBr2NO2
MW: 254.8600 g/mol
Melting Pt: 228…231 °C
Flash Pt: 120.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00185696
CAS Number: 1122-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR