3,4-Diaminobenzoic acid 97%

Supplier: Ambeed
Warning

A222285-25G A222285-5G A222285-1000G A222285-100G A222285-10G A222285-500G
103849-850EA 5 USD
103849-850 77088-454 77088-456 77192-920 77192-922 77192-924 77192-926
3,4-Diaminobenzoic acid 97%
3,4-Diaminobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 411.8 °C (760 mm Hg)
Melting Pt: 215…218 °C
Flash Pt: 202.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00007726
CAS Number: 619-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR