3,4-Bis(benzyloxy)phenol 97%

Supplier: Ambeed
Warning

A495236-100MG A495236-250MG A495236-1G A495236-5G
103828-250EA 125 USD
103828-250 103839-330 77143-988 77143-990 77307-596 77307-598
3,4-Bis(benzyloxy)phenol 97%
3,4-Bis(benzyloxy)phenol

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₃
MW: 306.36 g/mol
Boiling Pt: 490.4±35.0 °C (760 mm Hg)
Melting Pt: 110.5…111.5 °C
Flash Pt: 250.4 °C
Storage Temperature: Ambient
MDL Number: MFCD27952000
CAS Number: 27688-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR