3,4,7,8-Tetramethyl-1,10-phenanthroline 95%

Supplier: Ambeed
Warning

A145297-100G A145297-10G A145297-250MG A145297-25G A145297-1G A145297-5G
103803-518EA 8 USD
103803-518 103841-600 77272-716 77272-718 77272-720 77299-444 77316-884 77316-886
3,4,7,8-Tetramethyl-1,10-phenanthroline 95%
3,4,7,8-Tetramethyl-1,10-phenanthroline

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂
Boiling Pt: 435.2 °C (760 mm Hg)
Melting Pt: 277…280 °C
Flash Pt: 192.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00004974
CAS Number: 1660-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR