Tri-O-benzyl-D-glucal 97%

Supplier: Ambeed
Warning

Synonyms: 3,4,6-Tri-O-benzyl-D-glucal, 1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol 3,4,6-Tribenzyl Ether

A101138-10G A101138-250MG A101138-1G A101138-5G
103795-818EA 33 USD
103795-818 103821-506 76780-466 76780-468 77282-664 77282-666
Tri-O-benzyl-D-glucal 97%
Tri-O-benzyl-D-glucal

Precaución: for Research Use Only

Formula: C₂₇H₂₈O₄
Boiling Pt: 544.9 °C (760 mm Hg)
Melting Pt: 57…58 °C
Flash Pt: 129.7 °C
Storage Temperature: Freezer
MDL Number: MFCD00061640
CAS Number: 55628-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR