3,4,6-Trichloropyridazine 97%

Supplier: Ambeed
Warning

A110230-100G A110230-10G A110230-25G A110230-500G A110230-5G
103823-450EA 7 USD
103823-450 77270-834 77270-836 77270-838 77270-840 77283-962
3,4,6-Trichloropyridazine 97%
3,4,6-Trichloropyridazine

Precaución: for Research Use Only

Formula: C₄HCl₃N₂
MW: 183.4231 g/mol
Boiling Pt: 315.5 °C (760 mm Hg)
Flash Pt: 173.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00834957
CAS Number: 6082-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR