3,4,6-Tribromopyridazine 98%

Supplier: Ambeed
Warning

A211062-10G A211062-250MG
103818-074EA 35 USD
103818-074 103848-768 76873-678 76873-680
3,4,6-Tribromopyridazine 98%
3,4,6-Tribromopyridazine

Precaución: for Research Use Only

Formula: C₄HBr₃N₂
MW: 316.7761 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13193323
CAS Number: 55928-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR