3,4,5-Trimethoxyphenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,4,5-Trimethoxybenzeneboronic acid, Dihydroxy(3,4,5-trimethoxyphenyl)borane, (3,4,5-Trimethoxyphenyl)metaboric acid
A725512-100G
A725512-10G
A725512-250MG
A725512-1G
A725512-25G
A725512-5G
103840-764EA
41
USD
103840-764
76697-544
76697-546
76697-548
76844-980
76844-982
76844-984
3,4,5-Trimethoxyphenylboronic acid 98%
3,4,5-Trimethoxyphenylboronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₃BO₅ MW: 212.01 g/mol Boiling Pt: 370.6 °C (760 mm Hg) Melting Pt: 245 °C Flash Pt: 177.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075676 CAS Number: 182163-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...