(3,4,5-Trimethoxyphenyl)acetic acid 98%
Supplier: Ambeed
|
Warning
|
A138266-100G
A138266-25G
A138266-5G
A138266-500G
76798-124EA
87
USD
76798-124
77315-606
77315-608
77878-977
(3,4,5-Trimethoxyphenyl)acetic acid 98%
(3,4,5-Trimethoxyphenyl)acetic acid
Precaución: for Research Use Only
Formula:
(C₆H₂(OCH₃)₃)CH₂COOH Melting Pt: 117…120 °C Flash Pt: 138.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004336 CAS Number: 951-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...