3,4,5-Trihydroxybenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Gallic aldehyde, Pyrogallol-5-carboxaldehyde
A213223-1G
A213223-25G
A213223-5G
A213223-100MG
A213223-10G
A213223-250MG
A213223-100G
103818-268EA
30
USD
103818-268
103848-974
77087-458
77087-460
77087-462
77275-234
77275-236
77275-238
77748-328
3,4,5-Trihydroxybenzaldehyde 98%
3,4,5-Trihydroxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₆O₄ MW: 154.12 g/mol Boiling Pt: 355.3 °C (760 mm Hg) Flash Pt: 182.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003371 CAS Number: 13677-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...