3,4,5-Trifluorobenzoic acid 98%

Supplier: Ambeed
Warning

A612858-100G A612858-10G A612858-5G
103840-178EA 6 USD
103840-178 77117-388 77117-390 77307-948
3,4,5-Trifluorobenzoic acid 98%
3,4,5-Trifluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃F₃O₂
Melting Pt: 97…99 °C
Flash Pt: 105.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00074962
CAS Number: 121602-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR