3,4,5-Trifluoroaniline 98%

Supplier: Ambeed
Warning

A136437-100G A136437-1G A136437-25G
77285-530EA 65.92 USD
77285-530 77285-532 77315-192
3,4,5-Trifluoroaniline 98%
3,4,5-Trifluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 200.4 °C (760 mm Hg)
Melting Pt: 58…62 °C
Flash Pt: 82.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00083522
CAS Number: 163733-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR