trans-3,4,5-Trifluoro-4'-(4-n-pentylcyclohexyl)biphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 3,4,5-Trifluoro-4'-(trans-4-pentylcyclohexyl)-1,1'-biphenyl, 3,4,5-Trifluoro-4'-(trans-4-pentylcyclohexyl)biphenyl

A100125-1G A100125-5G
103795-262EA 8 USD
103795-262 103820-984 77282-436 77282-438
trans-3,4,5-Trifluoro-4'-(4-n-pentylcyclohexyl)biphenyl 97%
trans-3,4,5-Trifluoro-4'-(4-n-pentylcyclohexyl)biphenyl

Precaución: for Research Use Only

Formula: C₂₃H₂₇F₃
MW: 360.464 g/mol
Boiling Pt: 429.9 °C (760 mm Hg)
Flash Pt: 248.4 °C
Storage Temperature: Ambient
MDL Number: MFCD09839002
CAS Number: 137019-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR