3,4,5-Trichloropyridazine 97%

Supplier: Ambeed
Warning

A137781-10G A137781-1G
103802-506EA 7 USD
103802-506 77272-332 77298-926
3,4,5-Trichloropyridazine 97%
3,4,5-Trichloropyridazine

Precaución: for Research Use Only

Formula: C₄HCl₃N₂
MW: 183.42 g/mol
Melting Pt: 56…58 °C
Flash Pt: 178.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006464
CAS Number: 14161-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR