3,4,5-Trichlorobenzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3,4,5-Trichloro-benzoicacid

A175806-10G A175806-250MG A175806-1G
103807-088EA 34 USD
103807-088 103845-402 76784-430 76784-432 77082-750
3,4,5-Trichlorobenzoic acid 95%
3,4,5-Trichlorobenzoic acid

Precaución: for Research Use Only

Formula: C7H3Cl3O2
MW: 225.4600 g/mol
Melting Pt: 208…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00061128
CAS Number: 51-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR