3,4,5-Trichlorobenzaldehyde 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A109978-100MG A109978-5G A109978-250MG
103797-598EA 63 USD
103797-598 103823-406 77160-676 77310-214 77683-006
3,4,5-Trichlorobenzaldehyde 97%, Ambeed.Inc
3,4,5-Trichlorobenzaldehyde

Precaución: For research use only.

Formula: C₇H₃Cl₃O
MW: 209.45 g/mol
Boiling Pt: 280.5±35.0 °C (760 mm Hg)
Melting Pt: 90…91 °C
Flash Pt: 116.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18400105
CAS Number: 56961-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR