3,4,5-Trichloroaniline 98%

Supplier: Ambeed
Danger

A780434-25G A780434-1G A780434-5G
103840-976EA 19 USD
103840-976 77105-444 77308-318 77308-320
3,4,5-Trichloroaniline 98%
3,4,5-Trichloroaniline

Precaución: for Research Use Only

Formula: C₆H₄Cl₃N
MW: 196.46 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007769
CAS Number: 634-91-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR