3,4,5-Trichloro-2,6-dimethylpyridine 97%

Supplier: Ambeed
Warning

A100316-100MG A100316-250MG A100316-1G
103795-364EA 174 USD
103795-364 103821-066 77160-146 77160-148 77282-472
3,4,5-Trichloro-2,6-dimethylpyridine 97%
3,4,5-Trichloro-2,6-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₆Cl₃N
Boiling Pt: 240.0±35.0 °C (760 mm Hg)
Melting Pt: 78.3…79.9 °C
Flash Pt: 122.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00824780
CAS Number: 28597-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR