3,4,5-Tribromo-1H-pyrazole 98%
Supplier: Ambeed
|
Warning
|
A246666-100G
A246666-10G
A246666-25G
A246666-5G
103851-558EA
13
USD
103851-558
77000-592
77000-594
77090-856
77090-858
3,4,5-Tribromo-1H-pyrazole 98%
3,4,5-Tribromo-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₃HBr₃N₂ Boiling Pt: 381.5 °C (760 mm Hg) Melting Pt: 190…192 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00040248 CAS Number: 17635-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...