1,2-Diamino-3,4,5,6-tetrafluorobenzene 95%

Supplier: Ambeed
Warning

Synonyms: 3,4,5,6-Tetrafluorobenzene-1,2-diamine

A143724-100MG A143724-1G A143724-5G
103803-312EA 473 USD
103803-312 76984-304 77299-354 77299-356
1,2-Diamino-3,4,5,6-tetrafluorobenzene 95%
1,2-Diamino-3,4,5,6-tetrafluorobenzene

Precaución: for Research Use Only

Formula: C₆H₄F₄N₂
MW: 180.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00278787
CAS Number: 2993-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR