Tetrachloropyrocatechol 95%

Supplier: Ambeed
Danger

Synonyms: Tetrachlorobenzene-1,2-diol, 3,4,5,6-Tetrachloro-1,2-dihydroxybenzene, Tetrachlorocatechol, 3,4,5,6-Tetrachlorocatechol, 3,4,5,6-Tetrachlorobenzene-1,2-diol

A634333-250MG A634333-1G A634333-25G
103840-302EA 83 USD
103840-302 76790-802 77308-026 77602-600
Tetrachloropyrocatechol 95%
Tetrachloropyrocatechol

Precaución: for Research Use Only

Formula: C₆H₂Cl₄O₂
MW: 247.89 g/mol
Boiling Pt: 309.1 °C (760 mm Hg)
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00002190
CAS Number: 1198-55-6
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR