Piperonylonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-(Methylenedioxy)benzonitrile, 1,3-Benzodioxole-5-carbonitrile

A108908-100G A108908-10G A108908-25G A108908-1G A108908-5G
103797-404EA 5 USD
103797-404 103823-194 77270-730 77270-732 77270-734 77310-026 77310-028
Piperonylonitrile 98%
Piperonylonitrile

Precaución: for Research Use Only

Formula: C₈H₅NO₂
MW: 147.13 g/mol
Boiling Pt: 257.3 °C (760 mm Hg)
Melting Pt: 91…93 °C
Flash Pt: 115.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00005820
CAS Number: 4421-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR