3',4'-(Methylenedioxy)acetophenone 98%

Supplier: Ambeed
Warning

A407964-100G A407964-10G A407964-500G A407964-25G A407964-5G
103838-216EA 20 USD
103838-216 76694-816 76694-818 76694-820 76841-718 77307-120
3',4'-(Methylenedioxy)acetophenone 98%
3',4'-(Methylenedioxy)acetophenone

Precaución: for Research Use Only

Formula: C₉H₈O₃
MW: 164.16 g/mol
Boiling Pt: 294.6 °C (760 mm Hg)
Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00005831
CAS Number: 3162-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR