3,3-Dimethylglutaric acid 97%

Supplier: Ambeed
Warning

Synonyms: 3,3-Dimethylpentanedioic acid, 3,3-Dimethyl-1,5-pentanedioic acid

A230430-1G A230430-25G A230430-100G A230430-10G A230430-5G A230430-500G
103819-546EA 9 USD
103819-546 103850-446 77089-304 77089-306 77112-974 77112-976 77303-878 77685-630
3,3-Dimethylglutaric acid 97%
3,3-Dimethylglutaric acid

Precaución: for Research Use Only

Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 300.4 °C (760 mm Hg)
Melting Pt: 100…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00002716
CAS Number: 4839-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR