3,3-Dimethylindoline 97%

Supplier: Ambeed
Warning

A129561-100MG A129561-250MG
103801-302EA 62 USD
103801-302 76870-658 76870-660
3,3-Dimethylindoline 97%
3,3-Dimethylindoline

Precaución: for Research Use Only

Formula: C₁₀H₁₃N
Storage Temperature: Refrigerator
MDL Number: MFCD11847210
CAS Number: 1914-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR