3,3-Dimethylindolin-2-one 97%

Supplier: Ambeed
Warning

A288142-100MG A288142-10G A288142-250MG A288142-1G A288142-5G
103813-782EA 17 USD
103813-782 103834-924 76787-492 76787-494 76787-496 76837-474 76837-476
3,3-Dimethylindolin-2-one 97%
3,3-Dimethylindolin-2-one

Precaución: for Research Use Only

Formula: C10H11NO
MW: 161.2004 g/mol
Boiling Pt: 299.2 °C (760 mm Hg)
Flash Pt: 172.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00158549
CAS Number: 19155-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR