3,3-Dimethyl-2-oxobutyric acid 95%

Supplier: Ambeed
Danger

A519788-10G A519788-25G A519788-5G
103828-496EA 15 USD
103828-496 103839-604 76790-078 76843-608 77307-712
3,3-Dimethyl-2-oxobutyric acid 95%
3,3-Dimethyl-2-oxobutyric acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
Boiling Pt: 182.4 °C (760 mm Hg)
Melting Pt: 13 °C
Flash Pt: 78.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00154352
CAS Number: 815-17-8
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR