3,3'-Methylenediindole 98%

Supplier: Ambeed
Warning

A155014-100G A155014-100MG A155014-10G A155014-500G A155014-25G
103842-900EA 8 USD
103842-900 77286-242 77286-244 77286-246 77286-248 77318-636
3,3'-Methylenediindole 98%
3,3'-Methylenediindole

Precaución: for Research Use Only

Formula: C₁₇H₁₄N₂
Boiling Pt: 504.8 °C (760 mm Hg)
Melting Pt: 167 °C
Storage Temperature: Ambient
MDL Number: MFCD00195766
CAS Number: 1968-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR