3,3-Difluoropyrrolidine hydrochloride 97%

Supplier: Ambeed
Warning

A256205-10G A256205-250MG A256205-1G A256205-25G A256205-5G
103812-248EA 10 USD
103812-248 103852-112 76827-064 76827-066 77091-600 77091-602 77091-604
3,3-Difluoropyrrolidine hydrochloride 97%
3,3-Difluoropyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₇F₂N
Melting Pt: 133…136 °C
Storage Temperature: Ambient
MDL Number: MFCD03788948
CAS Number: 163457-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR