3,3-Difluorocyclobutanecarboxylic acid 98%

Supplier: Ambeed
Danger

A195512-1G A195512-25G A195512-5G A195512-10G
103816-758EA 15 USD
103816-758 103847-370 77085-318 77085-320 77085-322 77274-624
3,3-Difluorocyclobutanecarboxylic acid 98%
3,3-Difluorocyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₆F₂O₂
Boiling Pt: 204.6 °C (760 mm Hg)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD08705857
CAS Number: 107496-54-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR