3,3-Difluoroazetidine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A543812-1G
A543812-5G
A543812-100G
A543812-10G
A543812-250MG
A543812-25G
103828-650EA
18
USD
103828-650
103839-772
76843-826
76843-828
77279-944
77279-946
77279-948
77279-950
3,3-Difluoroazetidine hydrochloride 97%
3,3-Difluoroazetidine hydrochloride
Precaución: for Research Use Only
Formula:
C3H6ClF2N Melting Pt: 138…143 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05663714 CAS Number: 288315-03-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...