Propiolaldehyde diethyl acetal 97%

Supplier: Ambeed
Danger

Synonyms: Propargylaldehyde diethyl acetal, 3,3-Diethoxypropyne, Propioaldehyde diethyl acetal, Propynal diethyl acetal, 3,3-Diethoxy-1-propyne, 3,3-Diethoxyprop-1-yne

A684940-10G A684940-1G A684940-25G A684940-5G A684940-100G
103840-552EA 10 USD
103840-552 76727-604 76844-758 76844-760 76844-762 77602-740
Propiolaldehyde diethyl acetal 97%
Propiolaldehyde diethyl acetal

Precaución: for Research Use Only

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 38
Flash Pt: 31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009237
CAS Number: 10160-87-9
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR