3,3-Diethoxy-1-propanol 98%
Supplier: Ambeed
|
Warning
|
A382476-1G
A382476-25G
A382476-5G
A382476-10G
103826-716EA
37
USD
103826-716
103837-752
76841-068
76841-070
76841-072
76876-696
3,3-Diethoxy-1-propanol 98%
3,3-Diethoxy-1-propanol
Precaución: for Research Use Only
Formula:
C₇H₁₆O₃ Boiling Pt: 190.1 °C (760 mm Hg) Flash Pt: 91.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00074850 CAS Number: 16777-87-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...