3,3'-Dibromo-2,2'-bithiophene 98%

Supplier: Ambeed
Danger

A291231-1G A291231-25G A291231-5G A291231-100G A291231-100MG A291231-10G A291231-250MG
103813-898EA 10 USD
103813-898 103835-038 76837-622 76837-624 76837-626 77032-284 77032-286 77032-288 77032-290
3,3'-Dibromo-2,2'-bithiophene 98%
3,3'-Dibromo-2,2'-bithiophene

Precaución: for Research Use Only

Formula: C8H4Br2S2
MW: 324.0554 g/mol
Boiling Pt: 318.6 °C (760 mm Hg)
Melting Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00114806
CAS Number: 51751-44-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR